EMAIL THIS PAGE TO A FRIEND
FEBS letters
FANCD2 is a target for caspase 3 during DNA damage-induced apoptosis.
PMID 25176410
Abstract
The Fanconi anemia (FA) pathway, of which the FANCD2 protein is a key component, plays crucial roles in the maintenance of hematopoietic stem cells and suppression of carcinogenesis. However, the function of FANCD2 remains unclear. Here, we report that FANCD2 is a novel and specific substrate of caspase 3. Cleavage of FANCD2 by caspase 3 did not require either the FA core complex or mono-ubiquitylation of FANCD2, and was stimulated by p53. In addition, we identified the cleavage sites and generated cell lines that stably express a caspase-resistant FANCD2 mutant. Our data suggest that FANCD2 is regulated by caspase-mediated degradation during apoptosis induced by DNA damage.
Related Materials