EMAIL THIS PAGE TO A FRIEND
BioMed research international
A disintegrin and metalloproteinase 9 is involved in ectodomain shedding of receptor-binding cancer antigen expressed on SiSo cells.
PMID 25177692
Abstract
In several human malignancies, the expression of receptor-binding cancer antigen expressed on SiSo cells (RCAS1) is associated with aggressive characteristics and poor overall survival. RCAS1 alters the tumor microenvironment by inducing peripheral lymphocyte apoptosis and angiogenesis, while reducing the vimentin-positive cell population. Although proteolytic processing, referred to as "ectodomain shedding," is pivotal for induction of apoptosis by RCAS1, the proteases involved in RCAS1-dependent shedding remain unclear. Here we investigated proteases involved in RCAS1 shedding and the association between tumor protease expression and serum RCAS1 concentration in uterine cancer patients. A disintegrin and metalloproteinase (ADAM) 9 was shown to be involved in the ectodomain shedding of RCAS1. Given the significant correlation between tumor ADAM9 expression and serum RCAS1 concentration in both cervical and endometrial cancer as well as the role for ADAM9 in RCAS1 shedding, further exploration of the regulatory mechanisms by which ADAM9 converts membrane-anchored RCAS1 into its soluble form should aid the development of novel RCAS1-targeting therapeutic strategies to treat human malignancies.