EMAIL THIS PAGE TO A FRIEND
Environmental technology
Inactivation of dinoflagellate Scripsiella trochoidea in synthetic ballast water by advanced oxidation processes.
PMID 25182606
Abstract
Ship-borne ballast water contributes significantly to the transfer of non-indigenous species across aquatic environments. To reduce the risk of bio-invasion, ballast water should be treated before discharge. In this study, the efficiencies of several conventional and advanced oxidation processes were investigated for potential ballast water treatment, using a marine dinoflagellate species, Scripsiella trochoidea, as the indicator organism. A stable and consistent culture was obtained and treated by ultraviolet (UV) light, ozone (O3), hydrogen peroxide (H2O2), and their various combinations. UV apparently inactivated the cells after only 10 s of irradiation, but subsequently photo-reactivation of the cells was observed for all methods involving UV. O3 exhibited 100% inactivation efficiency after 5 min treatment, while H2O2 only achieved maximum 80% inactivation in the same duration. Combined methods, e.g. UV/O3 and UV/H2O2, were found to inhibit photo-reactivation and improve treatment efficiency to some degree, indicating the effectiveness of using combined treatment processes. The total residual oxidant (TRO) levels of the methods were determined, and the results indicated that UV and O3 generated the lowest and highest TRO, respectively. The synergic effect of combined processes on TRO generation was found to be insignificant, and thus UV/O3 was recommended as a potentially suitable treatment process for ballast water.