PloS one
The structural and functional organization of the podocyte filtration slits is regulated by Tjp1/ZO-1.
PMID 25184792
Abstract
Blood filtration in the kidney glomerulus is essential for physiological homeostasis. The filtration apparatus of the kidney glomerulus is composed of three distinct components: the fenestrated endothelial cells, the glomerular basement membrane, and interdigitating foot processes of podocytes that form the slit diaphragm. Recent studies have demonstrated that podocytes play a crucial role in blood filtration and in the pathogenesis of proteinuria and glomerular sclerosis; however, the molecular mechanisms that organize the podocyte filtration barrier are not fully understood. In this study, we suggest that tight junction protein 1 (Tjp1 or ZO-1), which is encoded by Tjp1 gene, plays an essential role in establishing the podocyte filtration barrier. The podocyte-specific deletion of Tjp1 down-regulated the expression of podocyte membrane proteins, impaired the interdigitation of the foot processes and the formation of the slit diaphragm, resulting in glomerular dysfunction. We found the possibility that podocyte filtration barrier requires the integration of two independent units, the pre-existing epithelial junction components and the newly synthesized podocyte-specific components, at the final stage in glomerular morphogenesis, for which Tjp1 is indispensable. Together with previous findings that Tjp1 expression was decreased in glomerular diseases in human and animal models, our results indicate that the suppression of Tjp1 could directly aggravate glomerular disorders, highlights Tjp1 as a potential therapeutic target.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA003267
Anti-CD2AP antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA003326
Anti-CD2AP antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2700911
Anti-CD2AP antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA023887
Anti-FYN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2100865
Anti-FYN antibody produced in rabbit, affinity isolated antibody, lyophilized powder
AV47047
Anti-NPHS2 antibody produced in rabbit, affinity isolated antibody, lyophilized powder
HPA049486
Anti-NPHS2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411415
Anti-NPHS2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
P0372
Anti-Podocin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA034631
Anti-SYNPO antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB3500585
Anti-SYNPO antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001636
Anti-TJP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102434
Anti-TJP1 antibody produced in rabbit, affinity isolated antibody
SAB2106392
Anti-TJP1 antibody produced in rabbit, affinity isolated antibody
SAB1302474
ANTI-TJP3 (N-TERM) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2102716
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
SAB2104329
Anti-WT1 antibody produced in rabbit, affinity isolated antibody
SAB1411399
Anti-WT1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution