EMAIL THIS PAGE TO A FRIEND
Journal of neurophysiology
Glycinergic inhibition to the medial nucleus of the trapezoid body shows prominent facilitation and can sustain high levels of ongoing activity.
PMID 25185813
Abstract
Neurons in the medial nucleus of the trapezoid body (MNTB) are well known for their prominent excitatory inputs, mediated by the calyx of Held. Less attention has been paid to the prominent inhibitory inputs that MNTB neurons also receive. Because of their auditory nature, both excitatory and inhibitory synapses are highly active in vivo. These high levels of activity are known to reduce excitatory synaptic currents considerably, such that in vivo synaptic currents produced by the calyx are smaller than typically measured in standard brain slice experiments. The goal of this study was to investigate the properties of the inhibitory inputs in the Mongolian gerbil (Meriones unguiculatus) under activity levels that correspond to those in the intact brain to facilitate a direct comparison between the two inputs. Our results suggest that inhibitory inputs to MNTB are largely mediated by a fast and phasic glycinergic component, and to a lesser degree by a GABAergic component. The glycinergic component can sustain prolonged high levels of activity. Even when challenged with stimulus patterns consisting of thousands of stimuli over tens of minutes, glycinergic inputs to MNTB maintain large conductances and fast decays and even facilitate substantially when the stimulation frequency is increased. The inhibition is mediated by a relatively small number of independent input fibers. The data presented here suggest that inhibitory inputs to MNTB sustain high levels of activity and need to be considered for a full understanding of mechanisms underlying processing of auditory information in MNTB.