PloS one
Histone H3 lysine 36 methyltransferase Whsc1 promotes the association of Runx2 and p300 in the activation of bone-related genes.
PMID 25188294
Abstract
The orchestration of histone modifiers is required to establish the epigenomic status that regulates gene expression during development. Whsc1 (Wolf-Hirschhorn Syndrome candidate 1), a histone H3 lysine 36 (H3K36) trimethyltransferase, is one of the major genes associated with Wolf-Hirshhorn syndrome, which is characterized by skeletal abnormalities. However, the role of Whsc1 in skeletal development remains unclear. Here, we show that Whsc1 regulates gene expression through Runt-related transcription factor (Runx) 2, a transcription factor central to bone development, and p300, a histone acetyltransferase, to promote bone differentiation. Whsc1-/- embryos exhibited defects in ossification in the occipital bone and sternum. Whsc1 knockdown in pre-osteoblast cells perturbed histone modification patterns in bone-related genes and led to defects in bone differentiation. Whsc1 increased the association of p300 with Runx2, activating the bone-related genes Osteopontin (Opn) and Collagen type Ia (Col1a1), and Whsc1 suppressed the overactivation of these genes via H3K36 trimethylation. Our results suggest that Whsc1 fine-tunes the expression of bone-related genes by acting as a modulator in balancing H3K36 trimethylation and histone acetylation. Our results provide novel insight into the mechanisms by which this histone methyltransferase regulates gene expression.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-866
Anti-acetyl-Histone H4 Antibody, 200 µL, Use Anti-acetyl-Histone H4 Antibody (Rabbit Polyclonal Antibody) has been published and validated in ChIP, WB, ICC to detect acetyl-Histone H4 also known as H4 histone family member A, histone 1-H4a.
SAB4503433
Anti-AML1 antibody produced in rabbit, affinity isolated antibody
SAB4503434
Anti-AML1 antibody produced in rabbit, affinity isolated antibody
SAB2102063
Anti-RUNX1 (ab1) antibody produced in rabbit, affinity isolated antibody
SAB2102064
Anti-RUNX1 (ab2) antibody produced in rabbit, affinity isolated antibody
R0406 Anti-RUNX1 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
SAB1412250
ANTI-RUNX1 antibody produced in mouse, clone 4E12, purified immunoglobulin, buffered aqueous solution
SAB1412470
ANTI-RUNX1 antibody produced in mouse, clone 3A8, purified immunoglobulin, buffered aqueous solution
SAB1412471
ANTI-RUNX1 antibody produced in mouse, clone 3A1, purified immunoglobulin, buffered aqueous solution
SAB1412488
ANTI-RUNX1 antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
HPA004176
Anti-RUNX1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV38073
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
SAB3500417
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB2104005
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
SAB2104009
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
SAB2104011
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
SAB2104913
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
SAB4300709
Anti-RUNX1 antibody produced in rabbit, affinity isolated antibody
AV36678
Anti-RUNX2 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV36679
Anti-RUNX2 (AB2) antibody produced in rabbit, IgG fraction of antiserum, lyophilized powder
SAB1412189
ANTI-RUNX2 antibody produced in mouse, clone 6E1, purified immunoglobulin, buffered aqueous solution
SAB1412247
ANTI-RUNX2 antibody produced in mouse, clone 4E5, ascites fluid, buffered aqueous solution
SAB1412248
ANTI-RUNX2 antibody produced in mouse, clone 4D7, purified immunoglobulin, buffered aqueous solution
SAB1412249
ANTI-RUNX2 antibody produced in mouse, clone 4B4, purified immunoglobulin, buffered aqueous solution
SAB1412665
ANTI-RUNX2 antibody produced in mouse, clone 2B9, purified immunoglobulin, buffered aqueous solution
SAB1412666
ANTI-RUNX2 antibody produced in mouse, clone 1D2, purified immunoglobulin, buffered aqueous solution
SAB1412667
ANTI-RUNX2 antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
SAB1412668
ANTI-RUNX2 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
HPA022040
Anti-RUNX2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106220
Anti-RUNX2 antibody produced in rabbit, affinity isolated antibody