EMAIL THIS PAGE TO A FRIEND
Nature communications
Resident CD4+ T cells accumulate in lymphoid organs after prolonged antigen exposure.
PMID 25189091
Abstract
Effector and memory CD4(+) T cells acquire distinct migratory properties depending on the type and location of the immune responses. Due to the highly dynamic nature of T cell circulation, the comprehensive analysis of these migratory routes requires dedicated experimental approaches. Here, we analyse the migration of effector/memory CD4(+) T cells by long-term in vivo cell tracking. We identify a resident population of antigen-experienced CD4(+) T cells that resides in lymph nodes and Peyer's patches without circulation or proliferation. Resident CD4(+) T cells constitute up to 50% of all effector/memory cells, including, but not limited to, follicular helper T cells. Furthermore, these non-circulating T cells possess a distinct T cell receptor repertoire and accumulate in Peyer's patches after continuous oral antigen exposure. Our results provide the first direct evidence for a resident population of effector/memory CD4(+) T cells that is retained in lymphoid tissues.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50553
α-tri-Calcium phosphate, Reagent for transient & stable DNA transfections
Ca3O8P2