EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Constituents of Vigna angularis and their in vitro anti-inflammatory activity.
PMID 25189119
Abstract
Nine non-phenolic compounds, including four furanylmethyl glycosides, angularides A-D, one ent-kaurane diterpene glycoside, angularin A, and four triterpenoid saponins, angulasaponins A-D, were isolated from seeds of Vigna angularis, together with eight known compounds. Their structures were elucidated on the basis of extensive 1D and 2D NMR spectroscopic analysis as well as chemical methods. Angularin A, angulasaponins A-C, and azukisaponins III and VI showed inhibition of nitric oxide production in LPS-activated RAW264.7 macrophages, with IC50 values ranging from 13μM to 24μM.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
XAD16
Amberlite® XAD16N, 20-60 mesh