EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
Galectin-1 mediates radiation-related lymphopenia and attenuates NSCLC radiation response.
PMID 25189484
Abstract
Radiotherapy can result in lymphopenia, which has been linked to poorer survival. Here, we test the hypothesis that radiotherapy-induced lymphopenia is mediated by a tumor-secreted factor, Galectin-1 (Gal-1), which possesses T-cell proapoptotic activities. Matched Gal-1 wild-type (WT) or null mice were implanted with Lewis lung carcinoma (LLC-1) that either expressed Gal-1 or had Gal-1 stably downregulated. Tumors were irradiated locally and circulating Gal-1 and T cells were measured. Tumor growth, lung metastasis, intratumoral T-cell apoptosis, and microvessel density count were quantified. Thiodigalactoside (TDG), a Gal-1 inhibitor, was used to inhibit Gal-1 function in another group of mice to validate the observations noted with Gal-1 downregulation. Lymphocyte counts, survival, and plasma Gal-1 were analyzed in cohorts of radiotherapy-treated lung [non-small cell lung cancer (NSCLC)] and head and neck cancer patients. LLC irradiation increased Gal-1 secretion and decreased circulating T cells in mice, regardless of host Gal-1 expression. Inhibition of tumor Gal-1 with either shRNA or thiodigalactoside ablated radiotherapy-induced lymphopenia. Irradiated shGal-1 tumors showed significantly less intratumoral CD8(+) T-cell apoptosis and microvessel density, which led to marked tumor growth delay and reduced lung metastasis compared with controls. Similar observations were made after thiodigalactoside treatment. Radiotherapy-induced lymphopenia was associated with poorer overall survival in patients with NSCLC treated with hypofractionated radiotherapy. Plasma Gal-1 increased whereas T-cell decreased after radiation in another group of patients. Radiotherapy-related systemic lymphopenia appeared to be mediated by radiotherapy-induced tumor Gal-1 secretion that could lead to tumor progression through intratumoral immune suppression and enhanced angiogenesis.