EMAIL THIS PAGE TO A FRIEND
Ultrastructural pathology
Ultrastructural morphologic changes in mycobacterial biofilm in different extreme condition.
PMID 25192360
Abstract
The aim of this study was to investigate the morphologic and ultrastructural features of biofilms of slow and fast-growing mycobacteria in different stress conditions, presence and absence of oleic acid albumin dextrose catalase (OADC) enrichment and at different temperatures: 30, 37 and 42 °C. Four hundred mycobacterial isolates were taken. The biomass of each biofilm was quantified using a modified microtiter plate assay method. Isolates were divided into those that formed fully established biofilms, moderately attached biofilms and weakly adherent biofilms by comparison with a known biofilm-forming strain. The large quantity of biofilm was produced by Mycobacterium smegmatis at temperature 37 and 42 °C as compared to 30 °C. Mycobacterium fortuitum and M. avium developed large amount of biofilm at 30 °C as compared to 37 and 42 °C. Mycobacterium tuberculosis developed strong biofilm at 37 °C and no biofilm at 30 and 42 °C in Sauton's media. The selected non-tuberculous mycobacteria and H37Rv developed strong biofilm in the presence of OADC enrichment in Sauton's medium. Microscopic examination of biofilms by scanning electron microscopy revealed that poorly adherent biofilm formers failed to colonize the entire surface of the microtiter well. While moderately adherent biofilm formers grew in uniform monolayers but failed to develop a mature three-dimensional structure. SEM analysis of an isolate representative of the group formed fully established biofilms with a textured, multi-layered, three-dimensional structure.