EMAIL THIS PAGE TO A FRIEND
Breast cancer research and treatment
JMJD2A-dependent silencing of Sp1 in advanced breast cancer promotes metastasis by downregulation of DIRAS3.
PMID 25193278
Abstract
Specificity protein 1(Sp1) is a ubiquitous transcription factor and is highly expressed in breast cancer. However, its expression pattern and role in breast cancer progression remain unclear. The purpose of this study is to examine the expression pattern of Sp1 and determine its role in breast cancer progression. Immunohistochemistry (IHC) was performed on breast cancer tissues to reveal the expression pattern of Sp1. Spearman rank correlation was used for clinical statistics. Gene and protein expressions were monitored by IHC analysis, quantitative polymerase chain reaction, and Western blot. Wound-healing and Transwell assays were conducted to assess the role of Sp1 in breast cancer. Co-immunoprecipitation, deletion mutagenesis, chromatin immunoprecipitation, and dual luciferase reporter gene assays were used for investigation of the regulatory network. Sp1 expression was downregulated in late stage breast cancer and in highly invasive breast cancer cell lines. Expression of Sp1 was negatively correlated with TNM staging (P = 0.002) and metastasis status (P = 0.023). Overexpression of Sp1 inhibited breast cancer cell migratory and invasive abilities, whereas knockdown of GTP-binding RAS-like 3 (DIRAS3, also known as ARHI, NOEY2) attenuated the inhibitory effects. Moreover, re-expression of DIRAS3 abolished Sp1 knockdown-mediated cell migration and invasion. Jumonji domain containing 2A (JMJD2A) inhibited Sp1 autoregulation and explains Sp1 expression pattern in breast cancer. Sp1 negatively regulated breast cancer metastasis by transcriptional activation of DIRAS3. Inhibition of Sp1 autoregulation by JMJD2A contributed to Sp1 expression pattern in breast cancer. Our findings provided evidence that targeted therapy against Sp1 might be useful in early stage breast cancer. However, in late stages, development of Sp1 activator may be more promising for breast cancer treatments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500095
Anti-JMJD2A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305107
ANTI-JMJD2A(N-TERMINAL) antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
HPA007610
Anti-KDM4A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2701287
Anti-KDM4A antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
AV37192
Anti-SP1 (AB1) antibody produced in rabbit, IgG fraction of antiserum
AV37193
Anti-SP1 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB2500981
Anti-SP1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1412220
ANTI-SP1 antibody produced in mouse, clone 4H6, purified immunoglobulin, buffered aqueous solution
SAB1412261
ANTI-SP1 antibody produced in mouse, clone 4B11, purified immunoglobulin, buffered aqueous solution
SAB1412262
ANTI-SP1 antibody produced in mouse, clone 2G5, purified immunoglobulin, buffered aqueous solution
SAB1412263
ANTI-SP1 antibody produced in mouse, clone 3H7, purified immunoglobulin, buffered aqueous solution
SAB1412264
ANTI-SP1 antibody produced in mouse, clone 1G9, purified immunoglobulin, buffered aqueous solution
S9809 Anti-Sp1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
HPA001853
Anti-SP1 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV43585
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
HPA012292
Anti-SP1 antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502836
Anti-SP1 antibody produced in rabbit, affinity isolated antibody
PLA0307
Goat anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
SAB5300072
Monoclonal Anti-KDM4A antibody produced in mouse, clone 5H1, ascites fluid
WH0006667M2
Monoclonal Anti-SP1 antibody produced in mouse, clone 1A5, purified immunoglobulin, buffered aqueous solution
SAB1404397
Monoclonal Anti-SP1 antibody produced in mouse, clone 4C8, purified immunoglobulin, buffered aqueous solution
PLA0044
Rabbit anti-Sp1 Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.