EMAIL THIS PAGE TO A FRIEND
Archives of microbiology
Jilinibacillus soli gen. nov., sp. nov., a novel member of the family Bacillaceae.
PMID 25193716
Abstract
A Gram-positive, aerobic, rod-shaped, motile, endospore-forming bacterium, designated strain A12(T), was isolated from a saline and alkali soil samples in Baicheng City, western of Jilin Province, China. Growth occurred in 15-45xa0°C (optimum, 30xa0°C) and at pH 7.0-11.5 (optimum, pH 9.0) and in the presence of 0-10xa0% (w/v) NaCl [optimum, 1-3xa0% (w/v) NaCl]. Meso-DAP was present in the peptidoglycan. The predominant menaquinone was MK-7. The major polar lipid profile was phosphatidylethanolamine, phosphatidylglycerol, phosphatidylinositol, phosphatidyl inositol-methyl and phosphotidylinositol dimannosid. The major fatty acid (>10xa0% of total fatty acids) was anteiso-C15:0. DNA Gxa0+xa0C content was 36.2xa0molxa0%. The level of 16S rRNA gene sequence similarity between strain A12(T) and other recognized species of the family was below 95.6xa0%. Phylogenetic analysis based on 16S rRNA gene sequence data indicated that the strain A12(T) fell with the family Bacillaceae and formed a distinct taxon. Based on physiological, chemotaxonomic and phylogenetic analyses, strain A12(T) was considered to represent a novel species of a new genus, for which the name Jilinibacillus soli gen. nov., sp. nov. was proposed. The type strain of Jilinibacillus soli was A12(T) (=GIMN1.014(T)xa0=xa0CCTCC M2011164(T)xa0=xa0KCTC 33417(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33240
2,6-Diaminopimelic acid, ≥97.0% (NT)
C7H14N2O4
D1377
2,6-Diaminopimelic acid, ≥98% (TLC)
C7H14N2O4