EMAIL THIS PAGE TO A FRIEND
Rheumatology (Oxford, England)
Bone resorption correlates with the frequency of CD5⁺ B cells in the blood of patients with rheumatoid arthritis.
PMID 25193807
Abstract
The prevention of bone resorption and subsequent joint destruction is one of the main challenges in the treatment of patients suffering from RA. Various mechanisms have previously been described that contribute to bone resorption in tightly defined cohorts. Here we analysed a cross-sectional cohort of RA patients and searched for humoral and cellular markers in the peripheral blood associated with bone resorption. We enrolled 61 consecutive RA patients positive for ACPA. Blood was analysed by flow cytometry to determine the percentages of regulatory T cells and B cell subpopulations. Cytokine (TNF-α, IL-6, IL-10) and ACPA levels as well as the bone resorption marker CTX-1 were determined from the patients' sera. Standard clinical disease parameters were included. Multivariate analyses showed that the percentages of CD5(+) B cells were positively correlated with CTX-1 serum levels. However, neither low-avidity ACPA nor serum IL-6 levels, both known to be produced by CD5(+) cells, were associated with CTX-1 in patients' sera. There was no correlation between CTX-1 levels and clinical parameters or ACPA levels. In summary, we found that the CD5(+) B cell population is associated with bone resorption as measured via serum CTX-1 levels in a cross-sectional cohort of RA patients. However, a possible functional link between CD5(+) B cells and bone resorption still needs to be defined.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42910
Diphenylcyclopropenone, purum, ≥98.0% (HPLC)
C15H10O
177377
Diphenylcyclopropenone, 98%
C15H10O