EMAIL THIS PAGE TO A FRIEND
Journal of trace elements in medicine and biology : organ of the Society for Minerals and Trace Elements (GMS)
Adipose tissue chromium and vanadium disbalance in high-fat fed Wistar rats.
PMID 25194956
Abstract
The primary objective of the current study is to investigate the relationship between adipose tissue chromium and vanadium content and adipose tissue dysfunction in a model of diet-induced obesity. A total of 26 female Wistar rats were fed either standard or high-fat diet (31.6% of fat from total caloric content) for 3 months. High-fat-feeding resulted in 21 and 33% decrease in adipose tissue chromium and vanadium content, respectively. No change was seen in hair chromium or vanadium levels. Statistical analysis revealed a significant inverse correlation of adipose tissue Cr and V with animal morphometric parameters and adipocyte size. Significant inverse dependence was observed between adipose tissue Cr and V and serum leptin and proinflammatory cytokines' levels. At the same time, adipose tissue Cr and V levels were characterized by positive correlation between serum adiponectin and adiponectin/leptin ratio. Adipose tissue Cr and V were inversely correlated (p<0.05) with insulin and homeostatic model assessment insulin resistance index (HOMA-IR) levels. Cr and V concentrations were not correlated with serum glucose in either high-fat fed or control rats; however, both serum glucose and HOMA-IR levels were significantly higher in high-fat fed, compared to control, rats. The results allow to hypothesize that impairment of adipose tissue Cr and V content plays a certain role in the development of adipose tissue endocrine dysfunction in obesity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
357162
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
V
357170
Vanadium, foil, thickness 0.25 mm, 99.7% trace metals basis
V
266205
Vanadium, foil, thickness 0.5 mm, 99.7% trace metals basis
V
262935
Vanadium, powder, −325 mesh, 99.5% trace metals basis
V
266175
Vanadium, rod, diam. 6.2 mm, 99.7% trace metals basis
V
262927
Vanadium, turnings, 99.7% trace metals basis
V
774073
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
V
774065
Vanadium, powder, -100 mesh, 99.9% trace metals basis
V
GF98785388
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF25800386
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
V
GF46548692
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF21697106
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF76957004
Vanadium, microfoil, disks, 25mm, thinness 0.25μm, specific density 140.4μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF23355996
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
V
GF69763977
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
V
GF75438445
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
V
GF84027269
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
V
GF25047503
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
V
GF39723239
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
V
GF29805341
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%
V
GF03408555
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
V
GF30937732
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
V
GF57656391
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
V
GF25668344
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
V
GF26068855
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
V
GF67217430
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
V
GF78288946
Vanadium, wire reel, 2m, diameter 0.1mm, as drawn, 99.8%
V
GF08542975
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
V
GF58606409
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
V
GF02179672
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
V