EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Stepping down therapy in COPD.
PMID 25196116
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001288
Salbutamol sulfate for system suitability, European Pharmacopoeia (EP) Reference Standard
C13H21NO3 · 0.5H2SO4