EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Expression of uncoupling protein-2 remains increased within hibernating myocardium despite successful coronary artery bypass grafting at 4xa0wk post-revascularization.
PMID 25199570
Abstract
We have previously shown that mitochondrial uncoupling protein-2 (UCP-2) is increased in a swine model of hibernating myocardium (HM). Although UCP-2 reduces oxidant stress, it can promote inefficiency of the electron transport chain. In this study, we tested whether UCP-2 remains increased in revascularized HM (RHM) after coronary artery bypass grafting (CABG). Seven swine underwent thoracotomy with placement of a constrictor on the left anterior descending artery (LAD). Twelve weeks later, a left internal mammary artery graft was placed on the distal LAD. Four weeks post-CABG, computed tomography angiography documented patent grafts and function. At the terminal study, blood flow to the LAD and remote territories were assessed during high dose dobutamine and mitochondria isolated from both regions for analysis. Comparisons were made to a group of swine with HM who underwent constrictor placement without bypass grafting (nxa0=xa04). During dobutamine infusion, RHM demonstrated lower blood flows (2.44xa0±xa00.23 versus 3.43xa0±xa00.30xa0mL/min/g; Pxa0
Related Materials