EMAIL THIS PAGE TO A FRIEND
Molecular and cellular endocrinology
2-Arachidonoylglycerol impairs human cytotrophoblast cells syncytialization: influence of endocannabinoid signalling in placental development.
PMID 25199616
Abstract
A balanced cytotrophoblast cell turnover is crucial for placental development and anomalies in this process associated with gestational diseases. The endocannabinoid system (ECS) has emerged as a new player in several biological processes. However, its influence during placental development is still unknown. We report here the expression of the endocannabinoid 2-arachidonoylglycerol (2-AG) main metabolic enzymes in human cytotrophoblasts and syncytiotrophoblast. We also showed that 2-AG induced a decrease in placental alkaline phosphatase activity, human chorionic gonadotropin secretion and Leptin mRNA levels. Moreover, 2-AG reduced glial cell missing 1 and syncytin-2 transcription and the number of nuclei in syncytium. These effects were mediated by cannabinoid receptors and may result from 2-AG inhibition of the cAMP/PKA signalling pathway. Our data suggest that 2-AG may interfere with the biochemical and morphological differentiation of human cytotrophoblasts, through a CB receptor-dependent mechanism, shedding light on a role for the ECS in placental development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
1048
4-Nitrophenol, spectrophotometric grade
C6H5NO3
35836
4-Nitrophenol, PESTANAL®, analytical standard
C6H5NO3
48549
4-Nitrophenol, Reference Material
C6H5NO3
PHR1150
4-NITROPHENOL (Acetaminophen Related Compound F)(Paracetamol Impurity F), Pharmaceutical Secondary Standard; Certified Reference Material
C6H5NO3
40056
4-Nitrophenol solution, certified reference material, 5000 μg/mL in methanol
C6H5NO3
43575
4-Nitrophenol solution, stock solution, for mass spectrometry
C6H5NO3
49053
4-Nitrophenol solution, working solution, for mass spectrometry
C6H5NO3
P2126
L-Phenylalanine, reagent grade, ≥98%
C9H11NO2
P5482
L-Phenylalanine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
C9H11NO2
P8740
L-Phenylalanine, PharmaGrade, Ajinomoto, EP, JP, USP, manufactured under appropriate GMP controls for Pharma or Biopharmaceutical production, suitable for cell culture
C9H11NO2
78019
L-Phenylalanine, BioUltra, ≥99.0% (NT)
C9H11NO2
40541
L-Phenylalanine, certified reference material, TraceCERT®
C9H11NO2
W358509
L-Phenylalanine, 99%, FCC
C9H11NO2
91331
L-Phenylalanine, analytical standard, for Nitrogen Determination According to Kjeldahl Method
C9H11NO2
PHR1100
L-Phenylalanine, Pharmaceutical Secondary Standard; Certified Reference Material
C9H11NO2
1530503
L-Phenylalanine, United States Pharmacopeia (USP) Reference Standard
C9H11NO2
88185
Tetrazole solution, for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile
CH2N4