EMAIL THIS PAGE TO A FRIEND
Nucleic acids research
Direct binding of the Alu binding protein dimer SRP9/14 to 40S ribosomal subunits promotes stress granule formation and is regulated by Alu RNA.
PMID 25200073
Abstract
Stress granules (SGs) are formed in response to stress, contain mRNAs, 40S ribosomal subunits, initiation factors, RNA-binding and signaling proteins, and promote cell survival. Our study describes a novel function of the protein heterodimer SRP9/14 and Alu RNA in SG formation and disassembly. In human cells, SRP9/14 exists assembled into SRP, bound to Alu RNA and as a free protein. SRP9/14, but not SRP, localizes to SGs following arsenite or hippuristanol treatment. Depletion of the protein decreases SG size and the number of SG-positive cells. Localization and function of SRP9/14 in SGs depend primarily on its ability to bind directly to the 40S subunit. Binding of SRP9/14 to 40S and Alu RNA is mutually exclusive indicating that the protein alone is bound to 40S in SGs and that Alu RNA might competitively regulate 40S binding. Indeed, by changing the effective Alu RNA concentration in the cell or by expressing an Alu RNA binding-defective protein we were able to influence SG formation and disassembly. Our findings suggest a model in which SRP9/14 binding to 40S promotes SG formation whereas the increase in cytoplasmic Alu RNA following stress promotes disassembly of SGs by disengaging SRP9/14 from 40S.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A8456
4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride, ≥97%
C8H10FNO2S · HCl
A1410
Actinomycin D, from Streptomyces sp., ~98% (HPLC)
C62H86N12O16
A9415
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
C62H86N12O16
A4262
Actinomycin D, from Streptomyces sp., ≥95% (HPLC)
C62H86N12O16
01815
Actinomycin D, for fluorescence, ≥90% (HPLC), from Streptomyces sp.
C62H86N12O16
76307
Pefabloc® SC, analytical standard
C8H10FNO2S · HCl
791733
Potassium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
C2H3KO2
P1190
Potassium acetate, for molecular biology, ≥99.0%
C2H3KO2
P5708
Potassium acetate, BioXtra, ≥99.0%
C2H3KO2
P1222
Potassium acetate, meets USP testing specifications
C2H3KO2
255785
Potassium acetate, 99.98% trace metals basis, ~100 ppm Rb, <50 ppm Cs
C2H3KO2
236497
Potassium acetate, ACS reagent, ≥99.0%
C2H3KO2
P1147
Potassium acetate, ReagentPlus®, ≥99.0%
C2H3KO2
60035
Potassium acetate, BioUltra, for molecular biology, ≥99.0% (NT)
C2H3KO2
32309
Potassium acetate, puriss. p.a., ≥99% (perchloric acid titration)
C2H3KO2
25059
Potassium acetate, puriss., meets analytical specification of Ph. Eur., BP, E261, 99-101%
C2H3KO2
W292001
Potassium acetate, 98%
C2H3KO2
95843
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
C2H3KO2
U0600000
Urea, European Pharmacopoeia (EP) Reference Standard
CH4N2O
1706698
Urea, United States Pharmacopeia (USP) Reference Standard
CH4N2O
56180
Urea, analytical standard
CH4N2O
51465
Urea, tested according to Ph.Eur.
CH4N2O
08582
Urea solution, for microbiology, 40% in H2O
CH4N2O