EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Vasculogenic and hematopoietic cellular progenitors are scattered within the prenatal mouse heart.
PMID 25201347
Abstract
Vasculogenesis and hematopoiesis are co-localized in the embryonic body, but precise phenotypes of the cells contributing to these processes are not defined. The aim of this study was to characterize phenotypic profiles and location of putative vasculogenic and hematopoietic cellular progenitors in the embryonic mouse heart. Confocal microscopy, as well as ultrastructural and stereomicroscopic analyses, was performed on immunohistochemical whole-mount-stained or sectioned hearts at stages 11.5-14 dpc. A FASC analysis was conducted to quantify putative vasculogenic and hematopoietic cells. We found subepicardial blood islands in the form of foci of accumulation of cells belonging to erythroblastic and megakaryocytic lineages at various stages of maturation, exhibiting phenotypes: GATA2(+)/CD41(+), GATA2(-)/CD41(+), GATA2(+)/CD71(-), GATA2(-)/CD71(+), Fli1(+)/CD71(+), Fli1(-)/CD71(+), with a majority of cells expressing the Ter119 antigen, but none of them expressing Flk1. The subepicardium and the outflow tract endothelium were recognized to be the areas where progenitor cells were scattered or adjoining the endothelial cells. These progenitor cells were characterized as possessing the following antigens: CD45(+)/Fli1(+), CD41(+)/Flk1(+), Flk1(+)/Fli1(+). A FACS analysis demonstrated that the CD41/Flk1 double-positive population of cells constituted 2.68% of total cell population isolated from 12.5 dpc hearts. Vessels and tubules were positive for CD31, Flk1, Fli1, Tie2, including blood islands endothelia. The endocardial wall endothelia were found to function as an anchoring apparatus for megakaryocytes releasing platelets into the cardiac cavities. Phenotypic characteristics of vasculogenic (Flk1(+)/Fli1(+)) and hematopoietic (GATA2(+)/CD71(+), CD41(+)/GATA2(+)) progenitors, as well as the putative hemogenic endothelium (Flk1(+)/CD41(+)) in embryonic mouse hearts, have been presented. Cardiac blood islands, the subepicardium and endothelium of the outflow tract cushions have been defined as areas where these progenitor cells can be found.