EMAIL THIS PAGE TO A FRIEND
eLife
A critical role for mTORC1 in erythropoiesis and anemia.
PMID 25201874
Abstract
Red blood cells (RBC) must coordinate their rate of growth and proliferation with the availability of nutrients, such as iron, but the signaling mechanisms that link the nutritional state to RBC growth are incompletely understood. We performed a screen for cell types that have high levels of signaling through mTORC1, a protein kinase that couples nutrient availability to cell growth. This screen revealed that reticulocytes show high levels of phosphorylated ribosomal protein S6, a downstream target of mTORC1. We found that mTORC1 activity in RBCs is regulated by dietary iron and that genetic activation or inhibition of mTORC1 results in macrocytic or microcytic anemia, respectively. Finally, ATP competitive mTOR inhibitors reduced RBC proliferation and were lethal after treatment with phenylhydrazine, an inducer of hemolysis. These results identify the mTORC1 pathway as a critical regulator of RBC growth and proliferation and establish that perturbations in this pathway result in anemia.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500210
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4500736
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4500737
Anti-4E-BP1 antibody produced in rabbit, affinity isolated antibody
SAB4300474
Anti-EIF4EBP1 (Ab-36) antibody produced in rabbit, affinity isolated antibody
SAB4300475
Anti-EIF4EBP1 (Ab-45) antibody produced in rabbit, affinity isolated antibody
HPA023501
Anti-EIF4EBP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2106955
Anti-EIF4EBP1 antibody produced in rabbit, affinity isolated antibody
SAB2701669
Anti-eno2 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1408571
Anti-ENO2 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1303638
ANTI-HBA2 (CENTER) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
SAB2101383
Anti-LRRC23 antibody produced in rabbit, affinity isolated antibody
HPA037766
Anti-LRRC23 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305394
ANTI-NSE (Y236) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305395
ANTI-NSE (Y25) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300698
Anti-NSE antibody produced in rabbit, affinity isolated antibody
SAB4300187
Anti-phospho-RPS6 (pSer235) antibody produced in rabbit, affinity isolated antibody
P26252
Phenylhydrazine, 97%
C6H8N2
PLA0118
Rabbit anti-Phospho RPS6 (S235/236) Antibody, Affinity Purified, Powered by Bethyl Laboratories, Inc.
390062
Thiazole Orange, Dye content ~90 %
C26H24N2O3S2