EMAIL THIS PAGE TO A FRIEND
Stem cells and development
microRNA-21 mediates stretch-induced osteogenic differentiation in human periodontal ligament stem cells.
PMID 25203845
Abstract
microRNAs (miRNAs) are short 20- to 22-nucleotide noncoding RNAs that negatively regulate the expression of target genes at the post-transcriptional level. The expression of specific miRNAs and their roles in the osteogenic differentiation of human periodontal ligament stem cells (PDLSCs) exposed to mechanical stretch remain unclear. Here, we found that stretch induced both osteogenic differentiation and the differential expression of miR-21 in PDLSCs. Furthermore, we identified activin receptor type IIB (ACVR2B) as a target gene of miR-21. Luciferase reporter assays showed that miR-21 interacts directly with the 3'-untranslated repeat sequence of ACVR2B mRNA. Mechanical stretch suppressed ACVR2B protein levels in PDLSCs, and this suppressive effect was modulated when endogenous miR-21 levels were either enhanced or inhibited. Both stretch and the expression of miR-21 altered endogenous ACVR2B protein levels and thus the osteogenic differentiation of PDLSCs. In addition, gain- and loss of function of ACVR2B mediated the osteogenic differentiation of PDLSCs. This study demonstrates that miR-21 is a mechanosensitive gene that plays an important role in the osteogenic differentiation of PDLSCs exposed to stretch.