EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Analysis of oxysterols and vitamin D metabolites in mouse brain and cell line samples by ultra-high-performance liquid chromatography-atmospheric pressure photoionization-mass spectrometry.
PMID 25204266
Abstract
We have developed an ultra-high-performance liquid chromatography-atmospheric pressure photoionization-tandem mass spectrometric (UHPLC-APPI-MS/MS) method for the simultaneous quantitative analyses of several oxysterols and vitamin D metabolites in mouse brain and cell line samples. An UHPLC-APPI-high resolution mass spectrometric (UHPLC-APPI-HRMS) method that uses a quadrupole-time of flight mass spectrometer was also developed for confirmatory analysis and for the identification of non-targeted oxysterols. Both methods showed good quantitative performance. Furthermore, APPI provides high ionization efficiency for determining oxysterols and vitamin D related compounds without the time consuming derivatization step needed in the conventionally used electrospray ionization method to achieve acceptable sensitivity. Several oxysterols were quantified in mouse brain and cell line samples. Additionally, 25-hydroxyvitamin D3 was detected in mouse brain samples for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H4014
25-Hydroxycholecalciferol, ≥98% (HPLC)
C27H44O2
17938
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
C27H44O2 · H2O
739669
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, 98% (CP)
C27H44O2
H-083
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material
C27H44O2
H-086
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material
C27H44O2
739642
25-Hydroxyvitamin D3 solution, 50 μg/mL in ethanol, 98% (CP)
C27H44O2
739650
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, 98% (CP)
C27H44O2
C2394
5-Cholesten-3β-ol-7-one, ≥90%
C27H44O2
30800
7-Dehydrocholesterol, ≥95.0% (HPLC)
C27H44O
C0166000
Calcifediol, European Pharmacopoeia (EP) Reference Standard
C27H44O2 · H2O
C9756
Cholecalciferol, ≥98% (HPLC)
C27H44O
C9774
Cholecalciferol, analytical standard
C27H44O
C1357
Cholecalciferol, meets USP testing specifications
C27H44O
C2100000
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
C27H44O
1131009
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
C27H44O
47763
Cholecalciferol (D3), analytical standard
C27H44O
PHR1237
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
C27H44O
C2150000
Cholecalciferol for system suitability, European Pharmacopoeia (EP) Reference Standard
C27H44O
D6513
Desmosterol, ≥84% (GC)
C27H44O
E5750
Ergocalciferol, 40,000,000 USP units/g
C28H44O
E0900000
Ergocalciferol, European Pharmacopoeia (EP) Reference Standard
C28H44O
1239005
Ergocalciferol, United States Pharmacopeia (USP) Reference Standard
C28H44O
95220
Ergocalciferol, ≥98.0% (sum of enantiomers, HPLC)
C28H44O
47768
Ergocalciferol (D2), analytical standard
C28H44O
PHR1238
Ergocalciferol (Vitamin D2), Pharmaceutical Secondary Standard; Certified Reference Material
C28H44O
PHR1317
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
89680
Toluene, analytical standard
C7H8
PHR1311
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
C7H8
740306
Vitamin D2 solution, 100 μg/mL in ethanol, 98% (CP)
C28H44O
V-024
Vitamin D2 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material
C28H44O