EMAIL THIS PAGE TO A FRIEND
Clinical cancer research : an official journal of the American Association for Cancer Research
mTOR inhibitors induce cell-cycle arrest and inhibit tumor growth in Epstein-Barr virus-associated T and natural killer cell lymphoma cells.
PMID 25208880
Abstract
Epstein-Barr virus (EBV) infects B cells, as well as T cells and natural killer (NK) cells, and is associated with T or NK cell lymphoid malignancies. In various tumor cells, mTOR performs an essential function together with Akt with regard to cell growth. We investigated the effects of mTOR inhibitors on EBV-associated T- and NK-cell lymphomas. We investigated the Akt/mTOR activation pathway in EBV-positive and -negative T- and NK-cell lines (SNT13, SNT16, Jurkat, SNK6, KAI3, and KHYG1). We evaluated the antitumor effects of mTOR inhibitors (rapamycin and its analogue, CCI-779) against these cell lines in culture and in a murine xenograft model that was established by subcutaneous injection of SNK6 cells into NOG mice. All EBV-positive and -negative T- and NK-cell lines tested displayed activation of the Akt/mTOR pathway, and treatment with mTOR inhibitors suppressed mTOR activation. The inhibitors induced G1 cell-cycle arrest and inhibited cell proliferation in T- and NK-cell lines. Overall, T cell lines were more sensitive to rapamycin, but there were no significant differences between EBV-positive and -negative cell lines. Treatment with rapamycin did not affect lytic or latent EBV gene expression. Intraperitoneal treatment with CCI-779 significantly inhibited the growth of established tumors in NOG mice and reduced the EBV load in peripheral blood. These results suggest that inhibition of mTOR signaling is a promising new strategy for improving treatment of EBV-associated T- and NK-cell lymphoma.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
37094
Rapamycin, VETRANAL, analytical standard
C51H79NO13
PZ0020
Temsirolimus, ≥98% (HPLC)
C56H87NO16