EMAIL THIS PAGE TO A FRIEND
Cellular oncology (Dordrecht)
BMP9 regulates cross-talk between breast cancer cells and bone marrow-derived mesenchymal stem cells.
PMID 25209393
Abstract
Breast cancer cells frequently metastasize to distant organs, including bone. Interactions between breast cancer cells and the bone microenvironment are known to enhance tumor growth and osteolytic damage. Here we investigated whether BMP9 (a secretary protein) may change the bone microenvironment and, by doing so, regulate the cross-talk between breast cancer cells and bone marrow-derived mesenchymal stem cells. After establishing a co-culture system composed of MDA-MB-231 breast cancer cells and HS-5 bone marrow-derived mesenchymal stem cells, and exposure of this system to BMP9 conditioned media, we assessed putative changes in migration and invasion capacities of MDA-MB-231 cells and concomitant changes in osteogenic marker expression in HS-5 cells and metastases-related genes in MDA-MB-231 cells. We found that BMP9 can inhibit the migration and invasion of MDA-MB-231 cells, and promote osteogenesis and proliferation of HS-5 cells, in the co-culture system. We also found that the BMP9-induced inhibition of migration and invasion of MDA-MB-231 cells may be caused by a decreased RANK ligand (RANKL) secretion by HS-5 cells, leading to a block in the AKT signaling pathway. From our data we conclude that BMP9 inhibits the migration and invasion of breast cancer cells, and promotes the osteoblastic differentiation and proliferation of bone marrow-derived mesenchymal stem cells by regulating cross-talk between these two types of cells through the RANK/RANKL signaling axis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB2100910
Anti-GDF2 antibody produced in rabbit, affinity isolated antibody
O1139 Anti-Osteoprotegerin antibody produced in goat, affinity isolated antibody, lyophilized powder
O7262 Anti-Osteoprotegerin antibody produced in goat, affinity isolated antibody
SAB1406202
Anti-TNFRSF11B antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA027013
Anti-TNFRSF11B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1410859
Anti-TNFRSF11B antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
SAB1409922
Anti-TNFSF11 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4503430
Anti-TNFSF11 antibody produced in rabbit, affinity isolated antibody
SAB1411542
Anti-TNFSF11 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
WH0002658M1
Monoclonal Anti-GDF2 antibody produced in mouse, clone 4D2, purified immunoglobulin, buffered aqueous solution
SAB5300009
Monoclonal Anti-TNFRSF11B antibody produced in mouse, clone 5G2, ascites fluid
SAB5300010
Monoclonal Anti-TNFRSF11B antibody produced in mouse, clone 5A11, ascites fluid
T2942 Monoclonal Anti-TRANCE antibody produced in mouse, clone 70525, purified immunoglobulin, lyophilized powder