EMAIL THIS PAGE TO A FRIEND
American journal of otolaryngology
Endothelial progenitor cells in patients with age-related hearing loss.
PMID 25212104
Abstract
This study was conducted to determine the concentration of EPCs in patients with ARHL. Twenty patients with ARHL were evaluated. The number of EPCs was analyzed by flow cytometry analysis of peripheral blood CD34(+)/CD133(+) cells. The concentration of circulating EPCs, both for CD34(+)/CD133(+) cells, was significantly lower in ARHL patients compared to controls (P<0.05). No statistically significant differences were found between these two groups in terms of the level of total cholesterol, LDL, HDL, triglycerides and GLU. The possible role of circulating epithelial progenitor cells in the pathogenesis of age related hearing loss should be considered based on their significant reduction in patients with ARHL, although the association alone does not prove causality. Further studies were warranted to confirm the role of circulating EPCs in the pathogenesis of ARHL.