EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Remote intrathecal morphine preconditioning confers cardioprotection via spinal cord nitric oxide/cyclic guanosine monophosphate/protein kinase G pathway.
PMID 25214258
Abstract
Remote intrathecal morphine preconditioning (RMPC) induces cardioprotection, but the underlying mechanisms of this effect is unknown. The aim of this study was to investigate the role of spinal cord nitric oxide/cyclic guanosine monophosphate/protein kinase G (NO/cGMP/PKG) signal pathway in the cardioprotection of RMPC in rats. Anesthetized, open chest, male Sprague-Dawley rats were assigned to one of eight treatment groups 3xa0d after intrathecal catheter placement. Before ischemia and reperfusion, RMPC received intrathecal morphine (3xa0μg/kg) by three cycles of 5-min infusions interspersed with 5-min infusion free periods. Intrathecally administrated a nonspecific nitric oxide synthase (NOS) inhibitor Nω-Nitro-L-arginine methyl ester (30xa0nmol), a specific guanylate cyclase inhibitor oxadiazolo [4,3-a] quinoxalin-1-one (11xa0nmol) and PKG inhibitor KT-5823 (20 pmol) 10xa0min before RMPC was used to evaluate the role of NO/cGMP/PKG of spinal cord. Ischemia and reperfusion injury were then induced by 30xa0min of left coronary artery occlusion, followed by 120xa0min of reperfusion. Infarct size, as a percentage of the area at risk, was determined by 2,3,5-triphenyltetrazolium staining. The content of cyclic guanosine monophosphate in the thoracic spinal cord was determined by radioimmunity protocol; the contents of nitric oxide and activity of NOS in the thoracic spinal cord were determined by nitrate reductase reduction and colorimetric methods; the expression of neuronal NOS (nNOS) and PKG in the thoracic spinal cord were determined by immunohistochemical and Western blotting method; the expression of nNOS messenger RNA was determined by reverse transcription-polymerase chain reaction method. RMPC group markedly reduced the infarct size compared with the control group. However, the cardioprotection of RMPC could be abolished by pretreatment with Nω-Nitro-L-arginine methyl ester, Oxadiazolo [4,3-a] quinoxalin-1-one, and KT-5823. RMPC enhanced nitric oxide , NOS, and cyclic guanosine monophosphate levels in the spinal cord. Meanwhile, RMPC increased PKG and nNOS protein or messenger RNA expression in the spinal cord. Spinal cord NO/cGMP/PKG signaling pathway mediates RMPC-induced cardioprotective effect.