EMAIL THIS PAGE TO A FRIEND
Journal of dental research
Spatiotemporally controlled microchannels of periodontal mimic scaffolds.
PMID 25216511
Abstract
Physiologic bioengineering of the oral, dental, and craniofacial complex requires optimized geometric organizations of fibrous connective tissues. A computer-designed, fiber-guiding scaffold has been developed to promote tooth-supporting periodontal tissue regeneration and functional restoration despite limited printing resolution for the manufacture of submicron-scaled features. Here, we demonstrate the use of directional freeze-casting techniques to control pore directional angulations and create mimicked topographies to alveolar crest, horizontal, oblique, and apical fibers of natural periodontal ligaments. For the differing anatomic positions, the gelatin displayed varying patterns of ice growth, determined via internal pore architectures. Regardless of the freezing coordinates, the longitudinal pore arrangements resulted in submicron-scaled diameters (~50 µm), along with corresponding high biomaterial porosity (~90%). Furthermore, the horizontal + coronal ([Formula: see text]) freezing orientation facilitated the creation of similar structures to major fibers in the periodontal ligament interface. This periodontal tissue-mimicking microenvironment is a potential tissue platform for the generation of naturally oriented ligamentous tissues consistent with periodontal ligament neogenesis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G9539
FreAlagin M gelatin, lyophilized powder
G9382
Gelatin from bovine skin, gel strength ~225 g Bloom, Type B
G9391 Gelatin from bovine skin, Type B, powder, BioReagent, suitable for cell culture
G6650
Gelatin from bovine skin, Type B
G7765 Gelatin from cold water fish skin, 40-50% in H2O
G7041 Gelatin from cold water fish skin, BioReagent, solid
G2500 Gelatin from porcine skin, gel strength 300, Type A
G6144
Gelatin from porcine skin, gel strength 90-110 g Bloom, Type A
G1890 Gelatin from porcine skin, powder, gel strength ~300 g Bloom, Type A, BioReagent, for electrophoresis, suitable for cell culture
G8150
Gelatin from porcine skin, for electrophoresis, Type A
G2625
Gelatin from porcine skin, gel strength ~175 g Bloom, Type A
G9136 Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture
04055 Gelatin from porcine skin, powder, for microbiology
48720 Gelatin from porcine skin, for microbiology, low gel strength
48722 Gelatin from porcine skin, medium gel strength, for microbiology
48724 Gelatin from porcine skin, high gel strength, for microbiology, for microbiology
39465 Gelatin from porcine skin, for microbiology, ultrahigh gel strength
G1393 Gelatin solution, Type B, 2% in H2O, tissue culture grade, sterile, BioReagent, suitable for cell culture
G0411 Prionex® Highly purified Type A, aqueous solution