EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
The cell death and DNA damages caused by the Tet-On regulating HSV-tk/GCV suicide gene system in MCF-7 cells.
PMID 25217394
Abstract
Ganciclovir (GCV) affects the molecular mechanism of cell death and DNA damage by the rAAV (recombinant adeno-associated virus)-mediated Tet-On/HSV-tk/GCV suicide gene system in human breast cancer cell line MCF-7. A rAAV/TRE/Tet-On/HSV-tk combining a Tet-On regulating system and a suicide gene HSV-tk was used to transfect human breast cancer cell line MCF-7, and therapeutic effects on this system were studied. Afterwards, we used RT-PCR, western blotting, and a modified comet-assay to explore the potential mechanism of the HSV-tk/GCV suicide gene system in breast cancer treatments. MTT assay has shown that the cell number of GCV+rAAV+Dox group was significantly decreased compared with that of other groups after treatment and flow cytometric analysis detected that there was a substantial increase of S phase cells in this group, which means the HSV-tk/GCV suicide gene system probably works on the cell cycle. RT-PCR detected the expression level of p21 increased and PCNA had an opposite trend. Western blotting detected the protein expression of p21 and p53 increased and PCNA, CDK1, cyclin B decreased in GCV+rAAV+Dox group. The modified comet-assay shown that the very small extra fragments generated by the GCV+rAAV+Dox group treatment are visible as a small cloud extending from the comet in the direction of electrophoresis. The therapeutic mechanism of the HSV-tk/GCV suicide gene system on human breast cancer cell line MCF-7 is probably by upregulating the expression of p21 through a p53-dependent DNA damage signalling pathway, leading the decrease of protein expression of PCNA, cyclin B, CDK1 in MCF-7 cells and promoting the cell cycle arrest at G1/S phase. In summary, the HSV-tk/GCV suicide gene system arouses the death of MCF-7 cells from blocking the cell cycle and DNA damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB4504499
Anti-phospho-p53 (pSer15) antibody produced in rabbit, affinity isolated antibody
SAB4503953
Anti-phospho-p53 (pSer20) antibody produced in rabbit, affinity isolated antibody
SAB4504500
Anti-phospho-p53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4504501
Anti-phospho-p53 (pSer33) antibody produced in rabbit, affinity isolated antibody
SAB4504592
Anti-phospho-p53 (pSer366) antibody produced in rabbit, affinity isolated antibody
SAB4504502
Anti-phospho-p53 (pSer37) antibody produced in rabbit, affinity isolated antibody
SAB4504593
Anti-phospho-p53 (pSer378) antibody produced in rabbit, affinity isolated antibody
P8982 Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution
SAB4503954
Anti-phospho-p53 (pSer392) antibody produced in rabbit, affinity isolated antibody
SAB4504503
Anti-phospho-p53 (pSer46) antibody produced in rabbit, affinity isolated antibody
SAB4504504
Anti-phospho-p53 (pSer6) antibody produced in rabbit, affinity isolated antibody
SAB4504505
Anti-phospho-p53 (pSer9) antibody produced in rabbit, affinity isolated antibody
SAB4504506
Anti-phospho-p53 (pThr18) antibody produced in rabbit, affinity isolated antibody
SAB4504591
Anti-phospho-p53 (pThr387) antibody produced in rabbit, affinity isolated antibody
SAB1306688 ANTI-PHOSPHO-P53(S20) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305629 ANTI-PHOSPHO-P53(S315) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306606
ANTI-PHOSPHO-P53(S9) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1306609 ANTI-PHOSPHO-P53(T18) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1305574 ANTI-PHOSPHO-P53(T55) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB4300073
Anti-phospho-TP53 (pSer15) antibody produced in rabbit, affinity isolated antibody
SAB4300079
Anti-phospho-TP53 (pSer315) antibody produced in rabbit, affinity isolated antibody
SAB4300076
Anti-phospho-TP53 (pSer33) antibody produced in rabbit, affinity isolated antibody
SAB4300077
Anti-phospho-TP53 (pSer37) antibody produced in rabbit, affinity isolated antibody
SAB4300078
Anti-phospho-TP53 (pSer46) antibody produced in rabbit, affinity isolated antibody
SAB4300072
Anti-phospho-TP53 (pSer6) antibody produced in rabbit, affinity isolated antibody
SAB4300074
Anti-phospho-TP53 (pThr18) antibody produced in rabbit, affinity isolated antibody