EMAIL THIS PAGE TO A FRIEND
The Journal of surgical research
Platelet-rich plasma-containing fragmin-protamine micro-nanoparticles promote epithelialization and angiogenesis in split-thickness skin graft donor sites.
PMID 25217513
Abstract
Platelet-rich plasma (PRP) contains multiple growth factors, and fragmin-protamine micro-nanoparticles (F-P M-NPs) significantly enhance and stabilize growth factors. The purpose of this study was to evaluate the effects of PRP-containing F-P M-NPs (PRP&F-P M-NPs) on wound repair in split-thickness skin graft (STSG-) donor sites (DS). A total of 56 inbred male rats were anesthetized and split-thickness skin graft donor site (STSG-DS) were created with a Padgett dermatome. PRP&F-P M-NPs, F-P M-NPs, PRP, and saline (control) were then intradermally injected evenly into the STSG-DSs. On 3, 4, 5, 7, and 10xa0d after creation of STSG-DS, skin sample sections were stained with hematoxylin and eosin to evaluate reepithelialization and angiogenesis. Treatment of STSG-DS with PRP&F-P M-NPs effectively promoted epithelialization and new vessel formation compared with those treated with PRP, F-P M-NPs, and control (saline). The intradermal injection of PRP&F-P M-NPs promotes epithelialization and angiogenesis in STSG-DS wounds.