EMAIL THIS PAGE TO A FRIEND
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie
Identification of local angiogenic and inflammatory markers in the menstrual blood of women with endometriosis.
PMID 25218120
Abstract
The aim of this study was to evaluate the presence of myeloperoxidase (MPO), N-acetyl-β-D-glucosaminidase (NAG), tumor necrosis factor alpha (TNF-α) and vascular endothelial growth factor (VEGF) in peripheral and menstrual blood in women with (n=10) and without (n=7) endometriosis. NAG and MPO activities were evaluated by enzymatic methods, whereas TNF-α and VEGF by immunoassay. No significant differences were found for these markers, neither in menstrual nor in peripheral blood between groups. Menstrual blood NAG (P=0.039) and MPO (P=0.0117) activities in the endometriosis group were significantly higher than in peripheral blood. NAG and MPO presented positive linear correlation in peripheral (P=0.07; r=0.641) and menstrual blood (P=0.01; r=0.603). These findings point to the existence of an increased local inflammatory activity in women with endometriosis.