EMAIL THIS PAGE TO A FRIEND
Experimental eye research
Time course of cytokine upregulation in the lacrimal gland and presence of autoantibodies in a predisposed mouse model of Sjögren's Syndrome: the influence of sex hormones and genetic background.
PMID 25218176
Abstract
Sjögren's Syndrome (SS) is a chronic, inflammatory autoimmune disease characterized by lacrimal gland lymphocytic infiltration and epithelial cell death, as well as by the presence of serum autoantibodies. Although the symptoms of this syndrome are well characterized, patients are not diagnosed until 5-10 years into disease progression; furthermore, the early series of events leading to the initiation of SS are not well understood. In order to better understand the early events of the disease, we have been using ovariectomized (OVX) NOD.B10.H2(b) mice as a genetically predisposed model of SS. Previously, we have shown that removal of ovarian hormones through ovariectomy accelerated the symptoms of this disease, and in early events of SS in the lacrimal glands, lymphocytic infiltration preceded acinar cell apoptosis. To further elucidate the earlier events of this disease in the SS animal model, we investigated the expression and concentration of pro-inflammatory cytokines in the lacrimal glands as well as the presence of autoantibodies in both lacrimal glands and serum. Six weeks old NOD.B10.H2(b) and C57BL/10 control mice were either sham-operated, OVX, OVX and treated with 17β-estradiol (E2), or OVX and treated with dihydrotestosterone (DHT). Lacrimal glands were collected at 3, 7, 21, and 30 days after surgery and analyzed for cytokines IL-1β, TNF-α, IFN-γ, IL-10, and IL-4 gene expression by using quantitative RT-PCR and for cytokine levels using ELISA. Furthermore, anti-Ro/SSA and anti-La/SSB autoantibodies were measured in the serum and lacrimal glands supernatants using ELISA. The results of this study showed that OVX caused a significant increase in the expression and levels of the cytokines IL-1β, TNF-α, and IL-4 in the lacrimal glands of the NOD.B10.H2(b) mice starting at 3 days after OVX, while a significant increase of IL-10 gene expression and levels was observed only at later experimental time points. A small but significant increase in the expression of IL-1β and IL-4 was observed only at later experimental time points in the lacrimal glands of OVX C57BL/10 mice, while no significant changes in the expression of TNF-α and IL-10 were seen at any experimental times in this group. No significant differences were observed in the levels of the cytokines IL-1β, TNF-α, IL-4, and IL-10 in the lacrimal glands of the OVX C57BL/10 mice at any of the experimental times studied compared to the sham-operated group. IFN-γ was not detected in either mouse strains at the level of mRNA and protein. OVX in the NOD.B10.H2(b) mice also caused an increase in the levels of anti-Ro/SSA autoantibodies in the serum only, while no anti-La/SSB autoantibodies were found in the serum or lacrimal gland supernatants. Physiological doses of E2 or DHT at time of OVX prevented the upregulation of cytokines and the presence of anti-Ro/SSA autoantibodies in these animals. These results showed that a decrease in the concentrations of ovarian hormones in the genetically predisposed mice accelerated the onset of the disease by upregulating various pro-inflammatory cytokines at different time points and promoting the formation of anti-Ro/SSA serum autoantibodies, creating an environment favorable for the initiation of SS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
HPA017287
Anti-SSB antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
AV40462
Anti-SSB antibody produced in rabbit, affinity isolated antibody
AV40461
Anti-SSB antibody produced in rabbit, IgG fraction of antiserum
AV38248
Anti-TRIM21 (AB2) antibody produced in rabbit, IgG fraction of antiserum
SAB1406479
Anti-TRIM21 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA005673
Anti-TRIM21 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB2102542
Anti-TRIM21 antibody produced in rabbit, affinity isolated antibody
RAB0582 Cotton Rat IL4 / Interleukin 4 ELISA Kit
RAB0298
Human IL-4 ELISA Kit, for serum, plasma, cell culture supernatant and urine
WH0006741M1
Monoclonal Anti-SSB antibody produced in mouse, clone 1D6-H5, purified immunoglobulin, buffered aqueous solution
SAB1404410
Monoclonal Anti-SSB antibody produced in mouse, clone 3H7-F7, purified immunoglobulin, buffered aqueous solution
SAB1402363
Monoclonal Anti-TRIM21 antibody produced in mouse, clone 2C9, purified immunoglobulin, buffered aqueous solution
RAB0299
Mouse IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0300
Mouse IL-4 ELISA Kit, for cell and tissue lysates
RAB0301
Rat IL-4 ELISA Kit, for serum, plasma and cell culture supernatant
RAB0302
Rat IL-4 ELISA Kit, for cell and tissue lysates