EMAIL THIS PAGE TO A FRIEND
European journal of obstetrics, gynecology, and reproductive biology
Determination of cathepsin G in endometrial tissue using a surface plasmon resonance imaging biosensor with tailored phosphonic inhibitor.
PMID 25218550
Abstract
Cathepsin G is a serine peptidase whose physiological role is mainly associated with an early immune response, anti-microbial activity as well as platelet activation or hydrolysis of coagulation factors. In addition, since the activity of cathepsin G has been associated with the development of various pathological disorders, the measurement of its activity in patient samples is of high interest. Unfortunately, the usefulness of common immunological methods is limited, since they cannot distinguish between catalytically active and inactive protease. Here we present the application of recently developed Surface Plasmon Resonance-based biosensor for the detection of active cathepsin G in human endometrium samples. The key element of the system is based on the irreversible binding of cathepsin G to its specific phosphonic-type inhibitor immobilized on the surface of the gold chip. The concentration of cathepsin G was measured in tissue samples from the group of patients with endometriosis as well as in the control group. The level of cathepsin G ascertained in endometrium tissue samples was over twice as high for the group of patients suffering from endometriosis as compared to the control group, with the median values of 0.5 pmol/mg and 0.2 pmol/mg, respectively. The SPR sensor armed with a specific irreversible phosphonic inhibitor represents a highly useful tool for the determination of catalytically active cathepsin G concentration in endometrial tissue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M9768
Cysteamine, ~95%
C2H7NS
30070
Cysteamine, ≥98.0% (RT)
C2H7NS