EMAIL THIS PAGE TO A FRIEND
Steroids
Synthesis and anti-cancer cell activity of pseudo-ginsenoside Rh2.
PMID 25218677
Abstract
β-d-Glucopyranoside,(3β,12β,20E)-12,25-dihydroxydammar-20(22)-en-3-yl (pseudo-ginsenoside Rh2) and its 20Z-isomer were synthesized from ginsenoside Rh2 under a mild condition, via a simple three-step called acetylation, elimination-addition and saponification. In addition, their activities were evaluated by eight different human tumor cells, compared with ginsenoside Rh2 group. Results indicated that the reaction in the side chain might greatly enhance the anti-proliferative activity of ginsenosides.