EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Dermatology
Erythrodermic CD8+ pseudolymphoma during infliximab treatment in a patient with psoriasis: use of cyclosporine as a rescue therapy.
PMID 25219741