PloS one
Blood pressure-lowering peptides from neo-fermented buckwheat sprouts: a new approach to estimating ACE-inhibitory activity.
PMID 25222748
Abstract
Neo-fermented buckwheat sprouts (neo-FBS) contain angiotensin-converting enzyme (ACE) inhibitors and vasodilators with blood pressure-lowering (BPL) properties in spontaneously hypertensive rats (SHRs). In this study, we investigated antihypertensive mechanisms of six BPL peptides isolated from neo-FBS (FBPs) by a vasorelaxation assay and conventional in vitro, in vivo, and a new ex vivo ACE inhibitory assays. Some FBPs demonstrated moderate endothelium-dependent vasorelaxation in SHR thoracic aorta and all FBPs mildly inhibited ACE in vitro. Orally administered FBPs strongly inhibited ACE in SHR tissues. To investigate detailed ACE-inhibitory mechanism of FBPs in living body tissues, we performed the ex vivo assay by using endothelium-denuded thoracic aorta rings isolated from SHRs, which demonstrated that FBPs at low concentration effectively inhibited ACE in thoracic aorta tissue and suppressed angiotensin II-mediated vasoconstriction directly associated with BPL. These results indicate that the main BPL mechanism of FBP was ACE inhibition in living body tissues, suggesting that high FBP's bioavailability including absorption, tissue affinity, and tissue accumulation was responsible for the superior ACE inhibition in vivo. We propose that our ex vivo assay is an efficient and reliable method for evaluating ACE-inhibitory mechanism responsible for BPL activity in vivo.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P8155
(R)-(−)-Phenylephrine hydrochloride, analytical standard
C9H13NO2 · HCl
A6625
Acetylcholine chloride, ≥99% (TLC)
C7H16ClNO2
A2661
Acetylcholine chloride, suitable for cell culture
C7H16ClNO2
A9101
Acetylcholine chloride, pkg of 150 mg (per vial)
C7H16ClNO2
1008501
Acetylcholine chloride, United States Pharmacopeia (USP) Reference Standard
C7H16ClNO2
Y0000002
Acetylcholine chloride, European Pharmacopoeia (EP) Reference Standard
C7H16ClNO2
C4042
Captopril, ≥98% (HPLC), powder
C9H15NO3S
C8856
Captopril, meets USP testing specifications
C9H15NO3S
C0430000
Captopril, European Pharmacopoeia (EP) Reference Standard
C9H15NO3S
1091200
Captopril, United States Pharmacopeia (USP) Reference Standard
C9H15NO3S
PHR1307
Captopril, Pharmaceutical Secondary Standard; Certified Reference Material
C9H15NO3S
Y0001182
Captopril for system suitability, European Pharmacopoeia (EP) Reference Standard
C9H15NO3S
P1250000
Phenylephrine hydrochloride, European Pharmacopoeia (EP) Reference Standard
C9H13NO2 · HCl
1533002
Phenylephrine hydrochloride, United States Pharmacopeia (USP) Reference Standard
C9H13NO2 · HCl
PHR1017
Phenylephrine Hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
C9H13NO2 · HCl
Y0000683
Phenylephrine hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
C9H13NO2 · HCl