EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
Analysis of structure-function relationships in cytochrome c oxidase and its biomimetic analogs via resonance Raman and surface enhanced resonance Raman spectroscopies.
PMID 25223590
Abstract
Cytochrome c oxidase (CcO) catalyzes the four electron reduction of molecular oxygen to water while avoiding the formation of toxic peroxide; a quality that is of high relevance for the development of oxygen-reducing catalysts. Resonance Raman spectroscopy has been used since many years as a technique to identify electron transfer pathways in cytochrome c oxidase and to identify the key intermediates in the catalytic cycle. This information can be compared to artificial systems such as modified heme-copper enzymes, molecular heme-copper catalysts or CcO/electrode complexes in order to shed light into the reaction mechanism of these non-natural systems. Understanding the structural commonalities and differences of CcO with its non-natural analogs is of great value for designing efficient oxygen-reducing catalysts. In this review therefore Raman spectroscopic measurements on artificial heme-copper enzymes and model complexes are summarized and compared to the natural enzyme cytochrome c oxidase. This article is part of a Special Issue entitled: Vibrational spectroscopies and bioenergetic systems.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BCR058
Continuous cast copper (O), BCR® certified Reference Material, rod
Cu
31284
Copper, electrolytic, Analytical Reagent, Reag. Ph. Eur., ≥99.9% (complexometric), wire
Cu
326445
Copper, bars (random sizes), ≥99.999% trace metals basis
Cu
254177
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Cu
326488
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Cu
266744
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Cu
349178
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Cu
349208
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Cu
12816
Copper, foil, ≥99.8% (complexometric)
Cu
203122
Copper, powder, 99.999% trace metals basis
Cu
266086
Copper, powder, <425 μm, 99.5% trace metals basis
Cu
207780
Copper, powder, <75 μm, 99%
Cu
326453
Copper, powder (spheroidal), 14-25 μm, 99%
Cu
357456
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Cu
264857
Copper, rod, diam. 19 mm, 99.9998% trace metals basis
Cu
365327
Copper, rod, diam. 11 mm, 99.9999% trace metals basis
Cu
223409
Copper, shot, −3-+14 mesh, 99%
Cu
520381
Copper, spherical shot, −20-+30 mesh, ≥99.5% trace metals basis
Cu
349224
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Cu
349216
Copper, wire, diam. 1.0 mm, ≥99.9%
Cu
326410
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Cu
326429
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Cu
61139
Copper, turnings, purum p.a., ≥99.0%
Cu
61141
Copper, purum p.a., ≥99.0%, powder
Cu
GF14058705
Copper, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.999%
Cu
GF71667366
Copper, wire reel, 1000m, diameter 0.04mm, annealed, 99.9%
Cu
GF21709650
Copper, wire reel, 1000m, diameter 0.04mm, hard, 99.9%
Cu
GF99677165
Copper, wire reel, 1000m, diameter 0.05mm, annealed, 99.9%
Cu
GF08494577
Copper, wire reel, 1000m, diameter 0.075mm, annealed, 99.9%
Cu
GF66965956
Copper, wire reel, 1000m, diameter 0.125mm, annealed, 99.9%
Cu