EMAIL THIS PAGE TO A FRIEND
Journal of cellular and molecular medicine
Neuronal porosome lipidome.
PMID 25224862
Abstract
Cup-shaped lipoprotein structures called porosomes are the universal secretory portals at the cell plasma membrane, where secretory vesicles transiently dock and fuse to release intravesicular contents. In neurons, porosomes measure ~15 nm and are comprised of nearly 40 proteins, among them SNAREs, ion channels, the G(αo) G-protein and several structural proteins. Earlier studies report the interaction of specific lipids and their influence on SNAREs, ion channels and G-protein function. Our own studies demonstrate the requirement of cholesterol for the maintenance of neuronal porosome integrity, and the influence of lipids on SNARE complex assembly. In this study, to further understand the role of lipids on porosome structure-function, the lipid composition of isolated neuronal porosome was determined using mass spectrometry. Using lipid-binding assays, the affinity of porosome-associated syntaxin-1A to various lipids was determined. Our mass spectrometry results demonstrate the presence of phosphatidylinositol phosphates (PIP's) and phosphatidic acid (PA) among other lipids, and the enriched presence of ceramide (Cer), lysophosphatidylinositol phosphates (LPIP) and diacylglycerol (DAG). Lipid binding assays demonstrate the binding of neuronal porosome to cardiolipin, and confirm its association with PIP's and PA. The ability of exogenous PA to alter protein-protein interaction and neurotransmitter release is further demonstrated from the study.