EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Ten-percent solar-to-fuel conversion with nonprecious materials.
PMID 25225379
Abstract
Direct solar-to-fuels conversion can be achieved by coupling a photovoltaic device with water-splitting catalysts. We demonstrate that a solar-to-fuels efficiency (SFE) > 10% can be achieved with nonprecious, low-cost, and commercially ready materials. We present a systems design of a modular photovoltaic (PV)-electrochemical device comprising a crystalline silicon PV minimodule and low-cost hydrogen-evolution reaction and oxygen-evolution reaction catalysts, without power electronics. This approach allows for facile optimization en route to addressing lower-cost devices relying on crystalline silicon at high SFEs for direct solar-to-fuels conversion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32900
Benalaxyl-M, PESTANAL®, analytical standard
C20H23NO3
B6768
Boric acid, BioReagent, for molecular biology, suitable for cell culture, suitable for plant cell culture, ≥99.5%
H3BO3
B7901
Boric acid, for electrophoresis, ≥99.5%
H3BO3
B2645
Boric acid, tablet, 1 g boric acid per tablet
H3BO3
B9645
Boric acid, cell culture tested, plant cell culture tested, ≥99.5%
H3BO3
B1934
Boric acid, Biotechnology Performance Certified, ≥99.5% (titration), Cell Culture Tested
H3BO3
202878
Boric acid, 99.999% trace metals basis
H3BO3
339067
Boric acid, 99.97% trace metals basis
H3BO3
B7660
Boric acid, BioXtra, ≥99.5%
H3BO3
B0394
Boric acid, ACS reagent, ≥99.5%
H3BO3
B0252
Boric acid, ReagentPlus®, ≥99.5%
H3BO3
15663
Boric acid, BioUltra, for molecular biology, ≥99.5% (T)
H3BO3
31146
Boric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, ≥99.8%
H3BO3
11607
Boric acid, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.5-100.5%, powder
H3BO3
426148
Boric acid-11B, ≥99 atom % 11B
H311BO3
225819
Hydrazine hydrate, reagent grade, N2H4 50-60 %
H4N2 · xH2O
53847
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
H4N2 · H2O
53855
Hydrazine hydrate solution, purum, ~25% in H2O
H4N2 · H2O
15622
Hydrazine hydrate solution, 24-26% (iodometric)
H4N2 · H2O
18412
Hydrazine hydrate solution, 78-82% (iodometric)
H4N2 · H2O
207942
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, 98%
H4N2 · H2O
49143
Mettler-Toledo Calibration substance ME 51143095, Potassium nitrate, traceable to primary standards (LGC)
KNO3
60377
Potassium hydroxide, tested according to Ph.Eur.
HKO
61699
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
HKO
00648
Potassium hydroxide solution, 28.5%, for flue-gas analysis
HKO
P8291
Potassium nitrate, BioReagent, suitable for cell culture, suitable for plant cell culture
KNO3
P6083
Potassium nitrate, BioXtra, ≥99.0%
KNO3
P6030
Potassium nitrate, cell culture tested
KNO3
542040
Potassium nitrate, 99.999% trace metals basis
KNO3
204110
Potassium nitrate, 99.99% trace metals basis
KNO3