EMAIL THIS PAGE TO A FRIEND
Journal of thoracic oncology : official publication of the International Association for the Study of Lung Cancer
Generation and sequencing of pulmonary carcinoid tumor cell lines.
PMID 25226425
Abstract
Pulmonary carcinoid tumors account for approximately 5% of all lung malignancies in adults, and comprise 30% of all carcinoid tumors. There are limited reagents available to study these rare tumors, and consequently no major advances have been made for patient treatment. We report the generation and characterization of human pulmonary carcinoid tumor cell lines to study underlying biology, and to provide models for testing novel chemotherapeutic agents. Tissue was harvested from three patients with primary pulmonary typical carcinoid tumors undergoing surgical resection. The tumor was dissociated and plated onto dishes in culture media. The established cell lines were characterized by immunohistochemistry, Western blotting, and cell proliferation assays. Tumorigenicity was confirmed by soft agar growth and the ability to form tumors in a mouse xenograft model. Exome and RNA sequencing of patient tumor samples and cell lines was performed using standard protocols. Three typical carcinoid tumor lines grew as adherent monolayers in vitro, expressed neuroendocrine markers consistent with the primary tumor, and formed colonies in soft agar. A single cell line produced lung tumors in nude mice after intravenous injection. Exome and RNA sequencing of this cell line showed lineage relationship with the primary tumor, and demonstrated mutations in a number of genes related to neuronal differentiation. Three human pulmonary typical carcinoid tumor cell lines have been generated and characterized as a tool for studying the biology and novel treatment approaches for these rare tumors.