EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Characterization of the polyphenol composition of 20 cultivars of cider, processing, and dessert apples (Malus × domestica Borkh.) grown in Virginia.
PMID 25228269
Abstract
Polyphenols and maturity parameters were determined in 20 apple cultivars with potential for hard cider production grown in Virginia, U.S.A. Concentrations of five classes of polyphenols were significantly different across cultivar for both peel and flesh. Total polyphenol concentration ranged from 0.9 μg/g wwb in flesh of Newtown Pippin to 453 μg/g wwb in peel of Red Delicious. Harrison, Granny Smith, Rome, Winesap, and Black Twig cultivars contained the highest concentration of total flavan-3-ols in flesh, indicating potential to impart desired astringency and bitterness to cider under processing conditions where extraction of polyphenols from peel is minimal. These results can inform selection of fruit juice, extracts, and byproducts for investigations of bioactivity and bioavailability of polyphenols, and provide baseline data for horticultural and processing research supporting the growing hard cider industry in Virginia. Based on these data, cultivars Harrison, Granny Smith, Rome, Winesap, and Black Twig show high potential for cider production in Virginia.