EMAIL THIS PAGE TO A FRIEND
Endocrine-related cancer
BRAF activates and physically interacts with PAK to regulate cell motility.
PMID 25228413
Abstract
Increased p21-activated kinase (PAK) signaling and expression have been identified in the invasive fronts of aggressive papillary thyroid cancers (PTCs), including those with RET/PTC, BRAFV600E, and mutant RAS expression. Functionally, thyroid cancer cell motility in vitro is dependent on group 1 PAKs, particularly PAK1. In this study, we hypothesize that BRAF, a central kinase in PTC tumorigenesis and invasion, regulates thyroid cancer cell motility in part through PAK activation. Using three well-characterized human thyroid cancer cell lines, we demonstrated in all cell lines that BRAF knockdown reduced PAK phosphorylation of direct downstream targets. In contrast, inhibition of MEK activity either pharmacologically or with siRNA did not reduce PAK activity, indicating MEK is dispensable for PAK activity. Inhibition of cell migration through BRAF loss is rescued by overexpression of either constitutive active MEK1 or PAK1, demonstrating that both signaling pathways are involved in BRAF-regulated cell motility. To further characterize BRAF-PAK signaling, immunofluorescence and immunoprecipitation demonstrated that both exogenously overexpressed and endogenous PAK1 and BRAF co-localize and physically interact, and that this interaction was enhanced in mitosis. Finally, we demonstrated that acute induction of BRAFV600E expression in vivo in murine thyroid glands results in increased PAK expression and activity confirming a positive signaling relationship in vivo. In conclusion, we have identified a signaling pathway in thyroid cancer cells which BRAF activates and physically interacts with PAK and regulates cell motility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SAB3500499
Anti-B-raf antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAB1412329
ANTI-BRAF antibody produced in mouse, clone 2E9, purified immunoglobulin, buffered aqueous solution
SAB1412330
ANTI-BRAF antibody produced in mouse, clone 2F7, purified immunoglobulin, buffered aqueous solution
SAB1412331
ANTI-BRAF antibody produced in mouse, clone 4G7, purified immunoglobulin, buffered aqueous solution
SAB1412332
ANTI-BRAF antibody produced in mouse, clone 3A6, purified immunoglobulin, buffered aqueous solution
SAB1412333
ANTI-BRAF antibody produced in mouse, clone 1F2, purified immunoglobulin, buffered aqueous solution
SAB1412334
ANTI-BRAF antibody produced in mouse, clone 3F5, purified immunoglobulin, buffered aqueous solution
SAB1412335
ANTI-BRAF antibody produced in mouse, clone 4F4, purified immunoglobulin, buffered aqueous solution
SAB1412336
ANTI-BRAF antibody produced in mouse, clone 1H7, purified immunoglobulin, buffered aqueous solution
SAB1412337
ANTI-BRAF antibody produced in mouse, clone 2F6, purified immunoglobulin, buffered aqueous solution
HPA001328
Anti-BRAF antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4300427
Anti-PAK1 (Ab-212) antibody produced in rabbit, affinity isolated antibody
SAB2500741
Anti-PAK1 antibody produced in goat, affinity isolated antibody, buffered aqueous solution
SAB1406219
Anti-PAK1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003565
Anti-PAK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB4502062
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502064
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502065
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB4502066
Anti-PAK1 antibody produced in rabbit, affinity isolated antibody
SAB1406298
Anti-PKN1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
HPA003982
Anti-PKN1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1411085
Anti-PKN1 antibody produced in rabbit, purified immunoglobulin, buffered aqueous solution
EHU127401 MISSION® esiRNA, esiRNA human BRAF (esiRNA1)
WH0000673M1
Monoclonal Anti-BRAF antibody produced in mouse, clone 3C6, ascites fluid, buffered aqueous solution
SAB1403617
Monoclonal Anti-BRAF antibody produced in mouse, clone 3D2, purified immunoglobulin, buffered aqueous solution
SAB5300503 Monoclonal Anti-BRAF antibody produced in mouse, clone 1H12, ascites fluid
WH0005058M1
Monoclonal Anti-PAK1 antibody produced in mouse, clone 1E11, purified immunoglobulin, buffered aqueous solution
SAB1400193
Monoclonal Anti-PAK1 antibody produced in mouse, clone 4D1, purified immunoglobulin, buffered aqueous solution
WH0005585M1
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1B10, purified immunoglobulin, buffered aqueous solution
SAB1404244
Monoclonal Anti-PKN1 antibody produced in mouse, clone 1A4, purified immunoglobulin, buffered aqueous solution