EMAIL THIS PAGE TO A FRIEND
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America
Chronic hepatitis B virus coinfection is associated with renal impairment among Zambian HIV-infected adults.
PMID 25228705
Abstract
Among 6789 HIV-infected Zambian adults screened for hepatitis B virus (HBV) coinfection, estimated glomerular filtration rate (eGFR) was 50-90 mL/minute/1.73 m(2) in 17.6% and <50 mL/minute/1.73 m(2) in 2.5%. Human immunodeficiency virus/HBV coinfection was associated with eGFR <50 mL/minute/1.73 m(2) (adjusted odds ratio, 1.96 [95% confidence interval, 1.34-2.86]), adjusted for age, sex, CD4(+) count, and World Health Organization disease stage.