EMAIL THIS PAGE TO A FRIEND
Journal of medicinal food
Composition and antiproliferative effect of essential oil of Origanum vulgare against tumor cell lines.
PMID 25230257
Abstract
Cancer is a leading cause of death and is responsible for one in eight deaths worldwide. The use of herbs as complementary medicine for cancer, especially advanced cancer, has recently increased. The aim of this study was to evaluate in vitro, the antiproliferative effect of Origanum vulgare against human breast adenocarcinoma (MCF-7), and human colon adenocarcinoma (HT-29). The essential oil (EO) was extracted from a bought amount of O. vulgare dried leaves and analyzed in a gas chromatograph interfaced with a mass selective detector. The cytotoxicity test was performed by sulforhodamine B assay. The results show that the EO is composed mostly of 4-terpineol and induces a high cytotoxicity effect in HT-29. In the MCF-7 cell line the EO was less effective. In conclusion, this study showed that O. vulgare main component is 4-terpineol and was effective in inducing cancer cell growth inhibition.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
74856
(−)-Linalool, analytical standard
C10H18O
62139
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
C10H18O
00020590
1,8-Cineole, primary pharmaceutical reference standard
C10H18O
01410590
Camphene, primary pharmaceutical reference standard
C10H16
442505
Camphene, analytical standard
C10H16
456055
Camphene, 95%
C10H16
Y0000176
Cineole, European Pharmacopoeia (EP) Reference Standard
C10H18O
W524905
Dipentene, mixture of isomers
C10H16
334111
Dipentene, technical, for use as solvent (for the paint industry), mixture of various terpenes
C10H16
42560
Dipentene, technical, for use as solvent (for the paint industry), mixture of various terpenes
C10H16
29210
Eucalyptol, analytical standard
C10H18O
95656
Eucalyptol, certified reference material, TraceCERT®
C10H18O
C80601
Eucalyptol, 99%
C10H18O
W246506
Eucalyptol, natural, ≥99%, FCC, FG
C10H18O
51782
Linalool, analytical standard
C10H18O
W263508
Linalool, ≥97%, FCC, FG
C10H18O
L2602
Linalool, 97%
C10H18O
64643
Myrcene, analytical standard
C10H16
04600585
Myrcene, primary pharmaceutical reference standard
C10H16
M100005
Myrcene, technical grade
C10H16
W276200
Myrcene, ≥95%, stabilized, FCC, FG
C10H16
30039
p-Cymene, analytical standard
C10H14
49679
p-Cymene, certified reference material, TraceCERT®
C10H14
C121452
p-Cymene, 99%
C10H14
W235601
p-Cymene, ≥97%, FG
C10H14
442732
p-Isopropyltoluene, analytical standard
C10H14
77663
Terpineol, mixture of isomers, analytical standard
C10H18O
86480
Terpineol, mixture of isomers, anhydrous
C10H18O
T3407
Terpineol, ~95%
C10H18O
W304514
Terpineol, natural
C10H18O