EMAIL THIS PAGE TO A FRIEND
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme
Life and death of human vascular endothelial cells by stimulation with free fatty acids through death receptors.
PMID 25230327
Abstract
Diabetic individuals may have elevated levels of serum free fatty acids and may exhibit injury to the vascular endothelial cells. This study was undertaken to determine the relationship between various free fatty acids (FFAs) and vascular endothelial cell injury and the molecular mechanisms linking FFA-induced vascular endothelial cells injury or protection. We observed the survival of HUVECs exposed to different FFAs, and our results revealed that the effects of various FFAs on the cell survival of HUVECs were significantly different. Palmitic acid (PA) markedly decreased the HUVEC survival rate in a time- and dose-dependent manner, but arachidonic acid (AA) significantly increased the cell survival rate and could partially prevent cellular apoptosis induced by PA. Interestingly, PA and AA could activate the same target receptor, TNF-R1. PA induced the apoptosis of HUVECs by initiating the death pathway (TNF-R1/TRADD/caspases 8 pathway), whereas AA enhanced cell survival to protect vascular endothelial cells by activating the survival pathway (TNF-R1/RIP/NF-κB 50/NF-κB 65).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M2003
MTT Formazan, powder
C18H17N5S
88417
MTT Formazan, MTT reduction end product
C18H17N5S