EMAIL THIS PAGE TO A FRIEND
American journal of neurodegenerative disease
Optineurin immunoreactivity in neuronal and glial intranuclear inclusions in adult-onset neuronal intranuclear inclusion disease.
PMID 25232514
Abstract
Optineurin (OPTN) is a multifunctional protein involved in cellular morphogenesis, vesicle trafficking, maintenance of the Golgi complex, and transcription activation through its interactions with the Rab8, myosin 6 (MYO6), huntingtin. Recently, OPTN immunoreactivity has been reported in intranuclear inclusions in patients with neuronal intranuclear inclusions disease (NIID). Other studies have shown that the RNA-binding protein, fused in sarcoma (FUS), is a component of intranuclear inclusions in NIID. We aimed to investigate the relationship between OPTN, its binding protein MYO6 and FUS in this study. In control subjects, OPTN (C-terminal) (OPTN-C) and MYO6 immunoreactivity was mainly demonstrated in the cytoplasm of neurons. In NIID patients, both neuronal intranuclear inclusions (NII) and glial intranuclear inclusions (GII) were immunopositive for MYO6 as well as OPTN-C. However, the intensity of OPTN-C immunostaining of the neuronal cytoplasm with and without NII was less than that of the control subjects. Double immunofluorescence staining for OPTN-C, ubiquitin (Ub), p62 and FUS revealed co-localization of these proteins within NII. Moreover, Ub positive inclusions were co-localized with MYO6. The percentage of co-localization of Ub with OPTN-C, FUS or MYO6 in NII was 100%, 52% and 92%, respectively. Ultrastructurally, the inclusions consisted of thin and thick filaments. Both filaments were immunopositive for Ub and OPTN-C. These findings suggest that OPTN plays a central role in the disease pathogenesis, and that OPTN may be a major component of NII.