EMAIL THIS PAGE TO A FRIEND
Organic letters
Nickel-catalyzed intramolecular [3 + 2 + 2] cycloadditions of alkylidenecyclopropanes. A straightforward entry to fused 6,7,5-tricyclic systems.
PMID 25232684
Abstract
A highly diastereo- and chemoselective intramolecular nickel-catalyzed cycloaddition of alkene- and alkyne-tethered alkynylidenecyclopropanes is reported. The method constitutes the first fully intramolecular [3 + 2 + 2] alkylidenecyclopropropane cycloaddition occurring via a proximal cleavage of the cyclopropane and makes it possible to build relevant 6,7,5-tricyclic frameworks in a single-pot reaction. Importantly, the reaction outcome is highly dependent on the characteristics of the nickel ligands.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF82156912
Nickel, rod, 100mm, diameter 12.0mm, 99%
Ni
GF11598194
Nickel, rod, 100mm, diameter 12.7mm, 99.98%
Ni
GF29576010
Nickel, rod, 100mm, diameter 16mm, 99.99+%
Ni
GF83584143
Nickel, rod, 100mm, diameter 2.0mm, 99.99+%
Ni
GF45193697
Nickel, rod, 100mm, diameter 2.0mm, annealed, 99%
Ni
GF11517948
Nickel, rod, 100mm, diameter 20.0mm, 99%
Ni
GF39205931
Nickel, rod, 100mm, diameter 25.4mm, 99.98%
Ni
GF69167920
Nickel, rod, 100mm, diameter 3.0mm, 99%
Ni
GF70332870
Nickel, rod, 100mm, diameter 3.3mm, 99.99+%
Ni
GF99799968
Nickel, rod, 100mm, diameter 30mm, 99%
Ni
GF90754241
Nickel, rod, 100mm, diameter 40mm, 99%
Ni
GF20561385
Nickel, rod, 100mm, diameter 5.0mm, 99.99+%
Ni
GF90932823
Nickel, rod, 100mm, diameter 6.35mm, 99%
Ni
GF50593002
Nickel, rod, 100mm, diameter 9.5mm, 99%
Ni
GF68355085
Nickel, rod, 200mm, diameter 1.5mm, 99.99+%
Ni
GF66250256
Nickel, rod, 200mm, diameter 10.0mm, 99.99+%
Ni
GF69613825
Nickel, rod, 200mm, diameter 12.0mm, 99%
Ni
GF49774034
Nickel, rod, 200mm, diameter 12.7mm, 99.98%
Ni
GF97332554
Nickel, rod, 200mm, diameter 16mm, 99.99+%
Ni
GF58660746
Nickel, rod, 200mm, diameter 2.0mm, 99.99+%
Ni
GF71556958
Nickel, rod, 200mm, diameter 2.0mm, annealed, 99%
Ni
GF39742577
Nickel, rod, 200mm, diameter 20.0mm, 99%
Ni
GF11244259
Nickel, rod, 200mm, diameter 25.4mm, 99.98%
Ni
GF51944665
Nickel, rod, 200mm, diameter 3.0mm, 99%
Ni
GF91609555
Nickel, rod, 200mm, diameter 3.3mm, 99.99+%
Ni
GF95866824
Nickel, rod, 200mm, diameter 30mm, 99%
Ni
GF44280974
Nickel, rod, 200mm, diameter 40mm, 99%
Ni
GF70172838
Nickel, rod, 200mm, diameter 5.0mm, 99.99+%
Ni
GF17475903
Nickel, rod, 200mm, diameter 6.35mm, 99%
Ni
GF24125071
Nickel, rod, 200mm, diameter 9.5mm, 99%
Ni