EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
A study of the effect of sequential injection of 5-androstenediol on irradiation-induced myelosuppression in mice.
PMID 25234002
Abstract
Herein, we aimed at examining the therapeutic effects of 5-androstenediol (5-AED), a natural hormone produced in the adrenal cortex, on radiation-induced myelosuppression in C3H/HeN mice. The mice were subjected to whole-body irradiation with a sublethal dose of 5 Gy gamma-irradiation to induce severe myelosuppression, and 5-AED (50 mg/kg) was administered subcutaneously. 5-AED was administrated 1 day before irradiation (pre-treatment) or twice weekly for 3 weeks starting from 1 h after irradiation (post-treatment). Treatment with 5-AED significantly ameliorated the decrease in the peripheral blood neutrophil and platelet populations in irradiated myelosuppressive mice, but had no effect on the lymphocyte population. It also ameliorated hypocellularity and disruption of bone marrow induced by irradiation and led to rapid recovery of myeloid cells. Further, it attenuated the decrease in spleen weight and megakaryocyte and myeloid cell populations in the spleen and promoted multilineage hematopoietic recovery. We found that a single injection of 5-AED produced only a temporary therapeutic effect, while sequential injection of 5-AED after irradiation had a more pronounced and prolonged therapeutic effect and reduced myelosuppression by irradiation. Thus, sequential injection of 5-AED after irradiation has therapeutic potential for radiation-induced myelosuppression when administered continuously and can be a significant therapeutic candidate for the management of acute radiation syndrome, particularly in a mass casualty scenario where rapid and economic intervention is important.