EMAIL THIS PAGE TO A FRIEND
Food chemistry
An improved UPLC method for the detection of undeclared horse meat addition by using myoglobin as molecular marker.
PMID 25236222
Abstract
In 2013, following the scandal of the presence of undeclared horse meat in various processed beef products across the Europe, several researches have been undertaken for the safety of consumer health. In this framework, an improved UPLC separation method has been developed to detect the presence of horse myoglobin in raw meat samples. The separation of both horse and beef myoglobins was achieved in only seven minutes. The methodology was improved by preparing mixtures with different composition percentages of horse and beef meat. By using myoglobin as marker, low amounts (0.50mg/0.50g, w/w; ∼0.1%) of horse meat can be detected and quantified in minced raw meat samples with high reproducibility and sensitivity, thus offering a valid alternative to conventional PCR techniques.