EMAIL THIS PAGE TO A FRIEND
Cancer gene therapy
MDA-7/IL-24 inhibits Nrf2-mediated antioxidant response through activation of p38 pathway and inhibition of ERK pathway involved in cancer cell apoptosis.
PMID 25236495
Abstract
Reactive oxygen species (ROS) have a crucial role in melanoma differentiation-associated gene-7 (MDA-7)/interleukin-24 (IL-24)-induced cancer cell apoptosis. However, cancer cell has a series of protective mechanisms to resist ROS damage. Nuclear factor erythroid 2-related factor 2 (Nrf2) activates antioxidant response element (ARE)-mediated gene expression involved in cellular protection against oxidative stress. As the Nrf2 repressor, Kelch-like ECH-associated protein-1 (Keap1) sequesters Nrf2 in cytoplasm to block Nrf2 nuclear translocation. In the present study, administration of MDA-7/IL-24 by means of tumor-selective replicating adenovirus (ZD55-IL-24) was used to investigate whether ZD55-IL-24 could attenuate Nrf2-mediated oxidative stress response in cancer cell. We found that ZD55-IL-24 effectively strengthened the association between Nrf2 and Keap1 to restrict Nrf2 nuclear translocation, thereby inhibiting ARE-dependent transcriptional response. To evaluate the detailed mechanism underlying the suppression of ZD55-IL-24 on Nrf2-mediated oxidative stress response, we further tested three different mitogen-activated protein kinase (MAPK) signaling pathways in A549 and HeLa cells transfected by ZD55-IL-24. Our data showed that ZD55-IL-24 inhibited extracellular signal-regulated kinase (ERK) signal pathway but activated p38 and c-Jun-NH2-kinase (JNK) signal pathways to exert the tumor-specific apoptosis. Moreover, ERK pathway inhibitor U0126 prevented Nrf2 phosphorylation at Ser40 to retard Nrf2 nuclear translocation, thus decreasing antioxidant gene transcription. In contrast, p38 pathway inhibitor SB203580 obviously promoted the dissociation of Nrf2 from Keap1 to promote antioxidant gene transcription. However, JNK pathway had no effect on Nrf2 subcellular localization or the association of Nrf2 with Keap1. Conclusively, our results indicate that ZD55-IL-24 inhibits Nrf2-mediated oxidative stress response not only by activating p38 signal pathway to potentiate the association of Nrf2 and Keap1 but also by suppressing ERK signal pathway to postpone Nrf2 nuclear translocation. Given the 'dark' side of Nrf2 on carcinoma cell survival and chemoresistance, our study provides a novel explanation about MDA-7/IL-24-induced cancer-specific apoptosis and therapeutic sensitization through suppression of the cytoprotective system.