EMAIL THIS PAGE TO A FRIEND
Experimental dermatology
Loss of cell invasiveness through PKC-mediated syndecan-1 downregulation in melanoma cells under anchorage independency.
PMID 25236603
Abstract
Anchorage-independent survival is one of the key features for malignant tumor cells. Whether specific gene alterations contributed by anchorage independency would further affect metastatic phenotypes of melanoma cells was unclear. We adapted suspension culture of melanoma cells to establish anchorage independency. The suspended melanoma cells lost their invasive abilities in vitro. Specific loss of laminin-binding ability in suspended melanoma cells was observed, which was correlated with downregulation of syndecan-1 as revealed by microarray and validated by qPCR and Western blot. Modulation of syndecan-1 expression level affected laminin binding, transwell migration and matrix metalloproteinase-2 secretion in melanoma cells. SDC1 expression and transwell migration were correlated with activity or level of protein kinase Cδ as evidence by specific inhibitors and shRNA transfection. In this study, we compared metastatic phenotypes and gene expressions of adherent and suspended melanoma cells. The anchorage independency led to protein kinase Cδ-mediated syndecan-1 downregulation, which contributed to loss of laminin-binding ability, reduced metalloproteinase-2 secretion and loss of invasiveness.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9786 Anti-Hepatocyte Growth Factor Receptor (c-Met) antibody produced in goat, affinity isolated antibody, lyophilized powder
H9911 Anti-Hepatocyte Growth Factor Receptor antibody produced in goat, lyophilized powder, affinity isolated antibody
SAB4300599
Anti-MET (Ab-1003) antibody produced in rabbit, affinity isolated antibody
SAB4300479
Anti-MET (Ab-1234) antibody produced in rabbit, affinity isolated antibody
SAB4300487
Anti-MET (Ab-1349) antibody produced in rabbit, affinity isolated antibody
SAB1406117
Anti-MET antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
SAB4501870
Anti-MET antibody produced in rabbit, affinity isolated antibody
AV40493
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
SAB2102036
Anti-RNMT antibody produced in rabbit, affinity isolated antibody
HPA039409
Anti-RNMT antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
HPA006185
Anti-SDC1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAB1305543 MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, IgG fraction of antiserum, buffered aqueous solution
SAB1305487
MONOCLONAL ANTI-CD138 antibody produced in mouse, clone 480CT5.4.3, crude ascites, buffered aqueous solution
WH0008731M1
Monoclonal Anti-RNMT antibody produced in mouse, clone 3H3-1D12, purified immunoglobulin, buffered aqueous solution